Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT5C3 Antibody (009), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290252APC
Description
NT5C3 Monoclonal antibody specifically detects NT5C3 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
NT5C3 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
51251 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
009 | |
Western Blot, Immunoprecipitation | |
5'-nucleotidase, cytosolic III, cN-IIIMGC87828, cytosolic 5'-nucleotidase 3, Cytosolic 5'-nucleotidase III, p36, P5N1, P5N-1, P5'N-1MGC27337, PN-IMGC87109, PSN1UMPH, Pyrimidine 5'-nucleotidase 1, UMPH1EC 3.1.3.5, uridine 5' monophosphate hydrolase 1, Uridine 5'-monophosphate hydrolase 1 | |
Rabbit immunized with purified, recombinant Human NT5C3 (Accession#: NP_001002010, Met12-Leu297) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction