Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NT5C3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101256
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101256 Supplier Invitrogen™ Supplier No. PA5101256
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NT5C3.

Pyrimidine 5-prime-nucleotidase (P5N), also called uridine 5-prime monophosphate hydrolase (UMPH), catalyzes the dephosphorylation of the pyrimidine 5-prime monophosphates UMP and CMP to the corresponding nucleosides. There are 2 isozymes of pyrimidine 5-prime nucleotidase in red blood cells, referred to as type I (UMPH1) and type II (UMPH2). The 2 enzymes are not separable by electrophoresis in humans but have distinct kinetic properties, and the proteins show no homology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT5C3A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NT5C3A
Gene Accession No. Q9D020, Q9H0P0
Gene Alias 1600024P05Rik; 2610206B05Rik; 3110004A18Rik; 5'-nucleotidase, cytosolic III; 5'-nucleotidase, cytosolic IIIA; 5'-nucleotidase, pyrimidine; 7-methylguanosine nucleotidase; 7-methylguanosine phosphate-specific 5'-nucleotidase; AI842776; cN-III; Cytosolic 5'-nucleotidase 3; cytosolic 5'-nucleotidase 3A; Cytosolic 5'-nucleotidase III; fb30h07; HSPC233; hUMP1; lupin; MGC27337; MGC87109; MGC87828; nt5c3; Nt5c3a; Nt5y; p36; P5N1; P5'N-1; P5N-1; PN-1; PN-I; POMP; PSN1; pyrimidine 5'-nucleotidase; pyrimidine 5'-nucleotidase 1; UMPH; UMPH1; Umph-1; Unknown (protein for MGC:133762); uridine 5'-monophosphate hydrolase 1; uridine monophosphatase 1; wu:fb30h07; zgc:66117
Gene Symbols Nt5c3, NT5C3A
Host Species Rabbit
Immunogen A synthesized peptide derived from human NT5C3A(Accession Q9H0P0), corresponding to amino acid residues A16-T66.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107569, 51251
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.