Learn More
Invitrogen™ NT5C3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552714
Description
Immunogen sequence: FDKLQQHSIP VFIFSAGIGD VLEEVIRQAG VYHPNVKVVS NFMDFDETGV LKGFKGELIH VFNKHDGALR NTEYFNQLKD NSNIILLGDS QGDLRMADGV ANVEHILKIG YLNDRVDELL EKYMDSYDIV LVQDESLEVA NSILQKI Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Pyrimidine 5-prime-nucleotidase (P5N), also called uridine 5-prime monophosphate hydrolase (UMPH), catalyzes the dephosphorylation of the pyrimidine 5-prime monophosphates UMP and CMP to the corresponding nucleosides. There are 2 isozymes of pyrimidine 5-prime nucleotidase in red blood cells, referred to as type I (UMPH1) and type II (UMPH2). The 2 enzymes are not separable by electrophoresis in humans but have distinct kinetic properties, and the proteins show no homology.
Specifications
NT5C3A | |
Polyclonal | |
Unconjugated | |
NT5C3A | |
1600024P05Rik; 2610206B05Rik; 3110004A18Rik; 5'-nucleotidase, cytosolic III; 5'-nucleotidase, cytosolic IIIA; 5'-nucleotidase, pyrimidine; 7-methylguanosine nucleotidase; 7-methylguanosine phosphate-specific 5'-nucleotidase; AI842776; cN-III; Cytosolic 5'-nucleotidase 3; cytosolic 5'-nucleotidase 3A; Cytosolic 5'-nucleotidase III; fb30h07; HSPC233; hUMP1; lupin; MGC27337; MGC87109; MGC87828; nt5c3; Nt5c3a; Nt5y; p36; P5N1; P5'N-1; P5N-1; PN-1; PN-I; POMP; PSN1; pyrimidine 5'-nucleotidase; pyrimidine 5'-nucleotidase 1; UMPH; UMPH1; Umph-1; Unknown (protein for MGC:133762); uridine 5'-monophosphate hydrolase 1; uridine monophosphatase 1; wu:fb30h07; zgc:66117 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51251 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H0P0 | |
NT5C3A | |
Recombinant protein corresponding to Human NT5C3A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.