Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NT5C3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563399 Supplier Invitrogen™ Supplier No. PA563399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IVSNYMDFNE DGFLQGFKGQ LIHTYNKNSS VCENCGYFQQ LEGKTNVILL GDSIG Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Specifically hydrolyzes 7-methylguanosine monophosphate (m(7)GMP) to 7-methylguanosine and inorganic phosphate (PubMed:23223233, PubMed:24603684). The specific activity for m(7)GMP may protect cells against undesired salvage of m(7)GMP and its incorporation into nucleic acids (PubMed:23223233). Also has weak activity for CMP (PubMed:23223233, PubMed:24603684). UMP and purine nucleotides are poor substrates (PubMed:23223233). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT5C3B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NT5C3B
Gene Accession No. Q969T7
Gene Alias 2610037D24Rik; 5'-nucleotidase, cytosolic IIIB; 5'-nucleotidase, cytosolic III-like; 7-methylguanosine nucleotidase; 7-methylguanosine phosphate-specific 5'-nucleotidase; AI841843; C330027I04Rik; cN-IIIB; cN-III-like protein; Cytosolic 5'-nucleotidase 3B; cytosolic 5'-nucleotidase III-like protein; cytosolic 5-nucleotidase III-like protein-like; N(7)-methylguanylate 5'-phosphatase; Nt5c3b; Nt5c3l
Gene Symbols NT5C3B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NT5C3B. Recombinant protein control fragment (Product #RP-103290).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115024
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.