Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NT5D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556447
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556447 Supplier Invitrogen™ Supplier No. PA556447
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GFFSHLPSQR PFRTLENDEE QEALPSLDKP GWYSQGNAVH LYELLKKMTG KPEPKVVYFG DSMHSDIFPA RHYSNWETVL ILEELR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

While the exact function of the protein encoded by this gene is not known, it belongs to the 5'(3')-deoxyribonucleotidase family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT5D1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NT5DC1
Gene Accession No. Q5TFE4
Gene Alias 5'-nucleotidase domain containing 1; 5'-nucleotidase domain-containing protein 1; 5'-nucleotidase, cytosolic II-like 1; 5'-nucleotidase, cytosolic II-like 1 protein; C6orf200; LP2642; Nt5c2l1; NT5DC1
Gene Symbols NT5DC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NT5D1. Recombinant protein control fragment (Product #RP-94960).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.