Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT5DC1 Antibody (OTI1E11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202914
Description
NT5DC1 Monoclonal specifically detects NT5DC1 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NT5DC1 | |
Monoclonal | |
0.56 mg/mL | |
Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
NT5DC1 | |
Full length human recombinant protein of human NT5DC1 (NP_689942) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
221294 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI1E11 | |
Unconjugated | |
5'-nucleotidase domain containing 1,5'-nucleotidase domain-containing protein 1, 5'-nucleotidase, cytosolic II-like 1, C6orf200, dJ486I3.1, LP2642, MGC131837, MGC24302,5'-nucleotidase, cytosolic II-like 1 protein, NT5C2L1 | |
Mouse | |
51.7 kDa | |
0.1 mL | |
Primary | |
Human, Rat, Canine, Primate, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction