Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT5DC1 Monoclonal Antibody (OTI3A6), TrueMAB™, OriGene
SDP

Catalog No. 501672702
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-2702 100 μL
50-254-2184 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-2702 Supplier OriGene Supplier No. TA501566
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

While the exact function of the protein encoded by this gene is not known, it belongs to the 5′(3′)-deoxyribonucleotidase family.

Specifications

Antigen NT5DC1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3A6
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NT5DC1
Gene Accession No. Q5TFE4
Gene Alias C6orf200, LP2642, NT5C2L1
Gene Symbols NT5DC1
Host Species Mouse
Immunogen Full length human recombit protein of human NT5DC1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221294, 475036
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.