Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NT5DC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542853 Supplier Invitrogen™ Supplier No. PA542853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IRKYDYNPSF AIRGLHYDIQ KSLLMKIDAF HYVQLGTAYR GLQPVPDEEV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT5DC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NT5DC2
Gene Accession No. Q9H857
Gene Alias 2510015F01Rik; 5'-nucleotidase domain containing 2; 5'-nucleotidase domain-containing protein 2; eye irido-corneal angle unknown protein; fb83e03; im:6907673; NT5DC2; RGD1305524; wu:fb83e03; wu:fe11h11; zgc:153357
Gene Symbols NT5DC2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human NT5DC2 (aa 107-156).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64943
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.