Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT5E, CD73 Monoclonal Antibody (1G5G8), CoraLite488, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc CL48867789100UL


This item is not returnable.
View return policy
Description
NT5E, CD73 Monoclonal antibody specifically detects NT5E, CD73 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin)Specifications
NT5E, CD73 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P42345, Q9JLN9 | |
Mouse | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1G5G8 | |
CoraLite 488 | |
MTOR | |
2210401F01Rik; 5 nucleotidase, ecto; 5' nucleotidase, ecto; 5'-NT; 5'-nucleotidase; 5'-nucleotidase ecto; 5'-nucleotidase, ecto (CD73); AI447961; CALJA; CD73; E5NT; ecto-5'-nucleotidase; eN; eNT; NT; Nt5; Nt5e; NTE; Purine 5-Prime-Nucleotidase | |
Protein G | |
RUO | |
2475, 56717 | |
Liquid | |
Human|Mouse |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction