Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT5E/CD73 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501727816 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7816 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7816 Supplier Proteintech Group Inc Supplier No. 122311AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD73 (Ecto-5-prime-nucleotidase, 5-prime-ribonucleotide phosphohydrolase) catalyzes the conversion at neutral pH of purine 5-prime mononucleotides to nucleosides, the preferred substrate being AMP. CD73 consists of a dimer of 2 identical 70 kDa subunits bound externally to the plasma membrane by a glycosyl phosphatidyl inositol linkage. CD73 is used as a marker of lymphocyte differentiation. Consequently, a deficiency of CD73 occurs in a variety of immunodeficiency diseases. Other forms of 5-prime nucleotidase exist in the cytoplasm and lysosomes and can be distinguished from CD73 by their substrate affinities, requirement for divalent magnesium ion, activation by ATP, and inhibition by inorganic phosphate. The CD73 gene has been localized to 6q14-q21 by immunofluorescence and a study of a panel of human x mouse hybrids that contained fragments of chromosome 6 as translocations. Defects in the CD73 gene can lead to the calcification of joints and arteries, and intestinal tuberculosis. Two transcript variants encoding different isoforms of CD73 have been found.

Specifications

Antigen NT5E/CD73
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NT5E
Gene Accession No. P21588, P21589, Q61503
Gene Alias 5 NT, 5 nucleotidase, 5′ nucleotidase, ecto (CD73), CD73, E5NT, Ecto 5 nucleotidase, eN, eNT, NT, NT5, NT5E, NTE
Gene Symbols NT5E
Host Species Rabbit
Immunogen NT5E/CD73 Fusion Protein Ag2869
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23959, 4907, 58813
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.