Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT5M Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | NT5M |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NT5M Polyclonal specifically detects NT5M in Human samples. It is validated for Western Blot.Specifications
NT5M | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
3'-nucleotidase, mitochondrialDNT2, 5', 5' nucleotidase, mitochondrial, 5(3)-deoxyribonucleotidase, Deoxy-5'-nucleotidase 2, dNT2, dNT-25'(3')-deoxyribonucleotidase, mitochondrial, EC 3.1.3, EC 3.1.3.-, mdN, mitochondrial 5' nucleotidase | |
NT5M | |
IgG | |
23 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q9NPB1 | |
56953 | |
Synthetic peptides corresponding to NT5M(5',3'-nucleotidase, mitochondrial) The peptide sequence was selected from the N terminal of NT5M. Peptide sequence ALRVLVDMDGVLADFEGGFLRKFRARFPDQPFIALEDRRGFWVSEQYGRL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title