Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NT5M Polyclonal Antibody

Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | NT5M |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5118920
![]() |
Invitrogen™
PA5118920 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121734
|
Invitrogen™
PA5121734 |
20 μL |
Each for $209.50
|
|
|||||
Description
NT5M Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Dephosphorylates specifically the 5' and 2'(3')- phosphates of uracil and thymine deoxyribonucleotides, and so protects mitochondrial DNA replication from excess dTTP. Has only marginal activity towards dIMP and dGMP.Specifications
NT5M | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse | |
NT5M | |
2010013E09Rik; 5' nucleotidase, mitochondrial; 5(3)-deoxyribonucleotidase; 5'(3')-deoxyribonucleotidase, mitochondrial; 5',3'-deoxyribonucleotidase, mitochondrial; 5',3'-nucleotidase, mitochondrial; AI846937; Deoxy-5'-nucleotidase 2; DNT2; dNT-2; mdN; mitochondrial 5' nucleotidase; NT5M | |
NT5M | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q8VCE6, Q9NPB1 | |
103850, 56953 | |
Recombinant protein 5'-Nucleotidase, Mitochondrial. The antigen corresponds to amino acid range 38-228 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title