Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NT5M Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560504
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560504 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560504 Supplier Invitrogen™ Supplier No. PA560504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FELEPLPGAV EAVKEMASLQ NTDVFICTSP IKMFKYCPYE KYAWVEKYFG PDFLEQIVLT RDKT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

Dephosphorylates specifically the 5' and 2'(3')- phosphates of uracil and thymine deoxyribonucleotides, and so protects mitochondrial DNA replication from excess dTTP. Has only marginal activity towards dIMP and dGMP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT5M
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NT5M
Gene Accession No. Q9NPB1
Gene Alias 2010013E09Rik; 5' nucleotidase, mitochondrial; 5(3)-deoxyribonucleotidase; 5'(3')-deoxyribonucleotidase, mitochondrial; 5',3'-deoxyribonucleotidase, mitochondrial; 5',3'-nucleotidase, mitochondrial; AI846937; Deoxy-5'-nucleotidase 2; DNT2; dNT-2; mdN; mitochondrial 5' nucleotidase; NT5M
Gene Symbols NT5M
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NT5M. Recombinant protein control fragment (Product #RP-100000).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56953
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.