Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT5M Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 207651AP150UL

Description
Dephosphorylates specifically the 5′ and 2′(3′)- phosphates of uracil and thymine deoxyribonucleotides, and so protects mitochondrial DNA replication from excess dTTP. Has only marginal activity towards dIMP and dGMP.Specifications
NT5M | |
Polyclonal | |
Unconjugated | |
NT5M | |
Deoxy 5 nucleotidase 2, dNT 2, dNT2, mdN, NT5M | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
103850, 287368, 56953 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VCE6, Q9NPB1 | |
Nt5m | |
NT5M Fusion Protein Ag14743 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction