Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTAN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531446 Supplier Invitrogen™ Supplier No. PA531446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Raji. Predicted reactivity: Mouse (94%), Rat (95%), Pig (93%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Side-chain deamidation of N-terminal asparagine residues to aspartate. Required for the ubiquitin-dependent turnover of intracellular proteins that initiate with Met-Asn. These proteins are acetylated on the retained initiator methionine and can subsequently be modified by the removal of N-acetyl methionine by acylaminoacid hydrolase (AAH). Conversion of the resulting N-terminal asparagine to aspartate by PNAD renders the protein susceptible to arginylation, polyubiquitination and degradation as specified by the N-end rule. This enzyme does not act on substrates with internal or C-terminal asparagines and does not act on glutamine residues in any position.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTAN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Ntan1
Gene Accession No. Q96AB6
Gene Alias asparagine-specific N-terminal amidase; NTAN1; N-terminal Asn amidase; N-terminal asparagine amidase; N-terminal asparagine amidohydrolase; PNAA; PNAD; Protein NH2-terminal asparagine amidohydrolase; protein NH2-terminal asparagine deamidase; protein N-terminal Asn amidase; protein N-terminal asparagine amidase; protein N-terminal asparagine amidohydrolase; protein NTN-amidase; RGD1305160
Gene Symbols Ntan1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NTAN1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123803
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.