Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NTB-A/SLAMF6/CD352 Antibody (040), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290508B
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Mouse samples. It is validated for Western Blot.Specifications
| NTB-A/SLAMF6/CD352 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| 40 | |
| Western Blot | |
| Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse NTB-A/SLAMF6/CD352 (NP_109635.1; Met1-Asn239). | |
| 0.1 mL | |
| Immunology | |
| 114836 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction