Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NTB-A/SLAMF6/CD352 Antibody (110), FITC

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290029F
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
NTB-A/SLAMF6/CD352 | |
Monoclonal | |
FITC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human SLAMF7 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
110 | |
ELISA, Sandwich ELISA Detection | |
Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human NTB-A/SLAMF6/CD352 (Q96DU3-1; Met1-Met226). | |
0.1 mL | |
Immunology | |
114836 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction