Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NTB-A/SLAMF6/CD352 Antibody (13G3-19D), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $560.50
Specifications
| Antigen | NTB-A/SLAMF6/CD352 |
|---|---|
| Clone | 13G3-19D |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14334403
![]() |
Novus Biologicals
NBP1433440.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NBP14334401
![]() |
Novus Biologicals
NBP1433440.1MG |
0.1 mg |
Each for $560.50
|
|
|||||
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.Specifications
| NTB-A/SLAMF6/CD352 | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 114836 | |
| The immunogen for this antibody was Ly108. | |
| Primary | |
| Store at 4C. Do not freeze. |
| 13G3-19D | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Mouse | |
| Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6 | |
| SLAMF6 | |
| IgG2a κ | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title