Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NTB-A/SLAMF6/CD352 Rabbit anti-Human, Clone: 105, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | NTB-A/SLAMF6/CD352 |
---|---|
Clone | 105 |
Dilution | Flow Cytometry 1:25-1:100 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry.Specifications
NTB-A/SLAMF6/CD352 | |
Flow Cytometry 1:25-1:100 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
114836 | |
IgG | |
Protein A purified |
105 | |
Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human | |
Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6 | |
Recombinant Human NTB-A/SLAMF6/CD352 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title