Learn More
Invitrogen™ NTCP Polyclonal Antibody

Description
NTCP Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | NTCP |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SLC10A1 |
| Gene Accession No. | Q14973 |
| Gene Alias | bile acid cotransporting polypeptide; cell growth-inhibiting gene 29 protein; GIG29; growth-inhibiting protein 29; hepatic sodium-dependent bile acid transporter; LOW QUALITY PROTEIN: sodium/bile acid cotransporter; MGC128766 protein; Na(+)/bile acid cotransporter; Na(+)/taurocholate transport protein; Na/taurocholate cotransporting polypeptide; NA-dependent cholate transporting protein; Ntcp; Ntcp1; SBACT; Slc10a1; sodium bile acid cotransporting polypeptide; sodium/bile acid cotransporter; sodium/tauro; sodium/taurocholate cotransporter; sodium/taurocholate cotransporting polypeptide; sodium-dependent bile acid cotransporter; sodium-dependent taurocholate cotransporting polypeptide; sodium-taurocholate cotransporting polypeptide; solute carrier family 10 (sodium/bile acid cotransporter family), member 1; solute carrier family 10 (sodium/bile acid cotransporter), member 1; solute carrier family 10 member 1; solute carrier family 10, member 1 |
| Gene Symbols | SLC10A1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.