Learn More
NTF2 Mouse anti-Human, Clone: 4H7F9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | NTF2 |
---|---|
Clone | 4H7F9 |
Concentration | 0.86 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6501
![]() ![]() |
Proteintech Group Inc
660631IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The MW of this protein is 14 kDa, and this antibody specifically recognizes the 14 kDa protein.
The protein encoded by this gene is a cytosolic factor that facilitates protein transport into the nucleus. It interacts with the nuclear pore complex glycoprotein p62. This encoded protein acts at a relative late stage of nuclear protein import, subsequent to the initial docking of nuclear import ligand at the nuclear envelope. It is thought to be part of a multicomponent system of cytosolic factors that assemble at the pore complex during nuclear import.Specifications
NTF2 | |
0.86 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NTF 2, NTF2, nuclear transport factor 2, NUTF 2, NUTF2, Placental protein 15, PP15 | |
NUTF2 | |
IgG1 | |
Protein G | |
Antibody |
4H7F9 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P61970 | |
10204 | |
NTF2 Fusion Protein Ag7879 | |
Primary | |
-20°C | |
NUTF2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.