Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534025
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534025 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534025 Supplier Invitrogen™ Supplier No. PA534025
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Rat, Hamster, Elephant, Panda, Dog, Bovine, Opossum (100%) Turkey (93%).

The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTF3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Ntf3
Gene Accession No. P18280, P20181, P20783, Q08DT3
Gene Alias AI316846; AI835689; HDNF; Nerve growth factor 2; neurotrophic factor; neurotrophin 3; neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF2; NGF-2; NT3; NT-3; NTF3; Ntf-3
Gene Symbols Ntf3
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from N-Terminus of human NTF3 / Neurotrophin 3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100774728, 18205, 486731, 4908, 532393, 81737
Target Species Human, Mouse, Rat, Canine, Monkey, Bovine, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.