Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech NTF4 Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P83G1MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P83G1MG 1 mg
500P83G50UG 50 μg
500P83G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P83G1MG Supplier PeproTech Supplier No. 500P83G1MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

AA Sequence of recombinant protein: GVSETAPASR RGELAVCDAV SGWVTDRRTA VDLRGREVEV LGEVPAAGGS PLRQYFFETR CKADNAEEGG PGAGGGGCRG VDRRHWVSEC KAKQSYVRAL TADAQGRVGW RWIRIDTACV CTLLSRTGRA. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human NT-4. Anti-Human NT-4-specific antibody was purified by affinity chromatography employing an immobilized Human NT-4 matrix. Sandwich ELISA: To detect Human NT-4 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human NT-4 (500-P83GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human NT-4. Western Blot: To detect Human NT-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human NT-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTF4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene NTF4
Gene Accession No. P34130
Gene Alias 2900040K06Rik; AI462899; GLC10; GLC1O; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 4; Neurotrophin 4/5; neurotrophin 5; Neurotrophin 5 (neurotrophin 4/5); neurotrophin 5 (neurotrophin 4/5); neurotrophin-4; Neurotrophin-5; neutrophic factor 4; Neutrophic factor 5; NT4; NT-4; NT4/5; NT-4/5; NT4/NT5; NT4P; NT5; NT-5; Ntf4; Ntf5; Ntf-5
Gene Symbols NTF4
Host Species Goat
Immunogen E.coli-derived Recombinant Human NT-4.
Purification Method Antigen affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4909
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.