Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670644
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-644 Supplier Invitrogen™ Supplier No. PA118392
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Avoid repetitive freeze/thaw cycles. Glycerol (1:1) may be added for an additional stability.

This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTF4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with no preservative
Gene NTF4
Gene Accession No. P34130, P34131, Q80VU4
Gene Alias 2900040K06Rik; AI462899; GLC10; GLC1O; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 4; Neurotrophin 4/5; neurotrophin 5; Neurotrophin 5 (neurotrophin 4/5); neurotrophin 5 (neurotrophin 4/5); neurotrophin-4; Neurotrophin-5; neutrophic factor 4; Neutrophic factor 5; NT4; NT-4; NT4/5; NT-4/5; NT4/NT5; NT4P; NT5; NT-5; Ntf4; Ntf5; Ntf-5
Gene Symbols NTF4, Ntf5
Host Species Sheep
Immunogen Recombinant human NT4.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25730, 4909, 78405
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.