Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NTH1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$181.90 - $487.50
Specifications
Antigen | NTH1 |
---|---|
Dilution | Western Blot 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:500 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100302SS
![]() |
Novus Biologicals
NB100302SS |
0.025 mL |
Each for $181.90
|
|
|||||
NB100302
![]() |
Novus Biologicals
NB100302 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
NTH1 Polyclonal specifically detects NTH1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NTH1 | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Base Excision Repair, Cancer, Chromatin Research, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
Whole antisera with 0.1% Sodium Azide | |
EC 4.2.99.18, endonuclease III-like protein 1, nth endonuclease III-like 1 (E. coli), OCTS3NTH1nth (E.coli endonuclease III)-like 1 | |
NTHL1 | |
IgG | |
Unpurified | |
37 kDa |
Western Blot 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:500 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
P78549 | |
4913 | |
Full-length recombinant human NTH1. [UniProt# P78549] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title