Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTHL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APPA6506
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APPA6506 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APPA6506 Supplier Invitrogen™ Supplier No. PA116506
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The target sequence has 93% sequence homology with bovine proteins. Suggested positive control: Hela whole cell lysate, K562 cell extracts, Hela cell extracts, K562 whole cell lysate.

Oxygen radicals damage chromosonal DNA causing cell death and inducing mutations. Although a major focus of oxidatively damaged DNA has centered on the repair of 8-oxo-G, a number of other damaged DNA sites are created by free-radical attack on DNA. The human NTH1 repair protein is one enzyme that has been shown to act on a large number of these other oxidatively damaged DNA sites.A homologue of E. coli Endonuclease III, NTH is a DNA glycosylase with apurinic/apyrimidinic lyase activity. The NTH protein has broad substrate specificity, including numerous ring saturation and fragmentation products of pyrimidines. Research indicates NTH plays a crucial role in removal of oxidative base lesions in mitochondrial DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTHL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene NTHL1
Gene Accession No. O35980, P78549
Gene Alias bifunctional DNA N-glycoslyase/DNA-(apurinic or apyrimidinic site) lyase; Bifunctional DNA N-glycosylase/DNA-(apurinic or apyrimidinic site) lyase; DNA glycoslyase/AP lyase; DNA glycosylase/AP lyase; Endonuclease III-like protein 1; FAP3; hNTH1; nth (endonuclease III)-like 1 (E.coli); nth endonuclease III-like 1; Nth1; NTHL 1; Nthl1; nth-like DNA glycosylase 1; OCTS 3; OCTS3; thymine glycol DNA glycosylase/AP lyase
Gene Symbols NTHL1
Host Species Rabbit
Immunogen A peptide derived from human NTH1, conjugated to KLH.
Purification Method Antigen affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18207, 29541, 4913
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.