Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nti-Integrin α5 chain (CD49e) Mouse anti-Human, RB780, Clone: IIA1, BD OptiBuild™

Catalog No. BDB755571 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB755571 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB755571 Supplier BD Biosciences Supplier No. 755571
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

nti-Integrin α5 chain (CD49e) Monoclonal antibody specifically detects nti-Integrin α5 chain (CD49e) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen nti-Integrin α5 chain (CD49e)
Applications Flow Cytometry
Classification Monoclonal
Clone IIA1
Concentration 0.2 mg/mL
Conjugate RB780
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Integrin α5; Integrin alpha-5; ITGA5; ITA5; FNRA; VLA-5; VLA5A
Host Species Mouse
Immunogen Human α5β1 Protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3678
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light.Do not freeze.The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.