Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nti-LOXL2, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABE48425UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE48425UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE48425UG Supplier MilliporeSigma Supplier No. ABE48425UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

nti-LOXL2, Cat. No. ABE484, is a highly specific rabbit polyclonal antibody that targets Lysyl oxidase homolog 2 and has been tested in Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

Lysyl oxidase homolog 2 (UniProt: Q9Y4K0; also known as EC: 1.4.3.13, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-1, LOXL2) is encoded by the LOXL2 gene (Gene ID: 4017) in human. LOXL2 localizes to both cytoplasmic and nuclear region. It catalyzes the post-translational oxidative deamination of lysine residues on target proteins leading to the formation of deaminated lysine (allysine). When secreted in extracellular matrix, it promotes cross-linking of extracellular matrix proteins by mediating oxidative deamination of peptidyl lysine residues in precursors to fibrous collagen and elastin. LOXL2 also acts as a regulator of sprouting angiogenesis, probably via collagen IV scaffolding. When present in the nucleus, it acts as a transcription corepressor and specifically mediates deamination of trimethylated Lysine 4 of histone H3 (H3K4me3), a specific tag for epigenetic transcriptional activation. Overexpression of LOXL2 has been reported in a number of cancers and its ability to promote epithelial to mesenchymal transition suggest that it might play a role in tumor progression. Higher expression has been correlated with metastasis and reduced survival in patients with aggressive breast cancer. LOXL2 activity is strongly induced in hypoxia and it is reported to be a direct transcriptional target of HIF1A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen nti-LOXL2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A 1:500 dilution from a representative lot detected LOXL2 in MDA-MB-231 cell lysate. Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected LOXL2 in human prostate and human placenta tissues. Immunoprecipitation Analysis: A representative lot detected LOXL2 in LoxL2 expressing vector (Courtesy Dr Sandra Peiro from the  Institut Hospital del Mar d'Investigacions Mèdiques (IMIM) in Barcelona Spain ).
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Lysyl oxidase homolog 2;EC: 1.4.3.13;Lysyl oxidase-like protein 2;Lysyl oxidase-related protein 2;Lysyl oxidase-related protein WS9-1
Gene Symbols LOXL2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal half of huamn Lysyl oxidase homolog 2.
Molecular Weight of Antigen ∽82 kDa obseerved; 86.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Lysyl oxidase homolog 2 in human cells. It targets an epitope within 16 amino acids from the N-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.