Learn More
nti-Rat/Mouse CD49a Hamster anti-Mouse, BUV563, Clone: Ha31/8, BD Biosciences™
Hamster Monoclonal Antibody
Supplier:  BD Biosciences 741306
Catalog No. BDB741306
Only null left
Add to Cart
Description
Description
nti-Rat/Mouse CD49a Monoclonal antibody specifically detects nti-Rat/Mouse CD49a in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
nti-Rat/Mouse CD49a
Monoclonal
Brilliant Ultraviolet 563
Itga1; Integrin alpha-1; Integrin α1; Laminin and collagen receptor; VLA-1a
Rat Emulsified Lewis Rat Glomeruli Cells
RUO
Mouse
IgG2 λ1
Flow Cytometry
Ha31/8
Aqueous buffered solution containing ≤0.09% sodium azide.
Hamster
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.