Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nti-Rat/Mouse CD49a Hamster anti-Mouse, BUV661, Clone: Ha31/8, BD Biosciences™

Catalog No. BDB741549 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB741549 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB741549 Supplier BD Biosciences Supplier No. 741549
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

nti-Rat/Mouse CD49a Monoclonal antibody specifically detects nti-Rat/Mouse CD49a in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen nti-Rat/Mouse CD49a
Applications Flow Cytometry
Classification Monoclonal
Clone Ha31/8
Conjugate Brilliant Ultraviolet 661
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Itga1; Integrin alpha-1; Integrin α1; Laminin and collagen receptor; VLA-1a
Host Species Hamster
Immunogen Rat Emulsified Lewis Rat Glomeruli Cells
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody.
Isotype IgG2 λ1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.