Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTMT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118127 Supplier Invitrogen™ Supplier No. PA5118127
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NTMT1 Polyclonal Antibody for Western Blot, IHC (P), IP

The METTL11A gene encodes an N-terminal methyltransferase for the RAN (MIM 601179) guanine nucleotide exchange factor regulator of chromosome condensation 1 (RCC1; MIM 179710). METTL11A enzyme alpha-N-methylates other protein targets such as SET (MIM 600960) and RB (MIM 180200).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTMT1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene NTMT1
Gene Accession No. Q9BV86
Gene Alias 2610205E22Rik; AD-003; AL033331; AL033332; Alpha N-terminal protein methyltransferase 1A; C9orf32; HOMT1A; methyltransferase like 11A; methyltransferase-like protein 11A; METTL11A; NRMT; NRMT1; N-terminal RCC1 methyltransferase; N-terminal Xaa-Pro-Lys N-methyltransferase 1; N-terminal Xaa-Pro-Lys N-methyltransferase 1, N-terminally processed; NTM1A; NTMT1; RGD1306582; X-Pro-Lys N-terminal protein methyltransferase 1A
Gene Symbols NTMT1
Host Species Rabbit
Immunogen E. coli-derived Human METTL11A/NTMT1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28989
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.