Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTMT1 Polyclonal Antibody

Catalog No. PIPA535925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535925 Supplier Invitrogen™ Supplier No. PA535925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (94%), Rat (93%), Pig (92%), Chicken (82%), Rhesus Monkey (100%), Bovine (91%).

The METTL11A gene encodes an N-terminal methyltransferase for the RAN (MIM 601179) guanine nucleotide exchange factor regulator of chromosome condensation 1 (RCC1; MIM 179710). METTL11A enzyme alpha-N-methylates other protein targets such as SET (MIM 600960) and RB (MIM 180200).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTMT1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene NTMT1
Gene Accession No. Q9BV86
Gene Alias 2610205E22Rik; AD-003; AL033331; AL033332; Alpha N-terminal protein methyltransferase 1A; C9orf32; HOMT1A; methyltransferase like 11A; methyltransferase-like protein 11A; METTL11A; NRMT; NRMT1; N-terminal RCC1 methyltransferase; N-terminal Xaa-Pro-Lys N-methyltransferase 1; N-terminal Xaa-Pro-Lys N-methyltransferase 1, N-terminally processed; NTM1A; NTMT1; RGD1306582; X-Pro-Lys N-terminal protein methyltransferase 1A
Gene Symbols NTMT1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human METTL11A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28989
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.