Learn More
Invitrogen™ NTN1 Polyclonal Antibody

Description
A suggested positive control for this product is MDCKII cell lysate.
Specifications
Specifications
| Antigen | NTN1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | NTN1 |
| Gene Accession No. | O09118, O95631, Q924Z9 |
| Gene Alias | AI561871; BC019633; Epididymis tissue protein Li 131P; HGNC:8029; netrin 1; netrin 1, mouse, homolog of; Netrin1; netrin-1; Ntn1; NTN1L |
| Gene Symbols | NTN1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.