Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675846
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-846 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-846 Supplier Invitrogen™ Supplier No. PA112866
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for this product is MDCKII cell lysate.

Netrin-1 is a protein target that plays a crucial role in various biological processes, particularly in the development and guidance of neuronal cells. It belongs to the netrin family of axon guidance molecules and is involved in mediating axon guidance, cell migration, and tissue morphogenesis during embryonic development. Netrin-1 functions by binding to specific receptors on the cell surface and providing directional cues for cellular movement and connectivity. Dysregulation of Netrin-1 has been implicated in various diseases, including cancer, neurological disorders, and cardiovascular conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTN1
Applications Western Blot
Classification Polyclonal
Concentration 0.094 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene NTN1
Gene Accession No. O09118, O95631, Q924Z9
Gene Alias AI561871; BC019633; Epididymis tissue protein Li 131P; HGNC:8029; netrin 1; netrin 1, mouse, homolog of; Netrin1; netrin-1; Ntn1; NTN1L
Gene Symbols NTN1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human Netrin 1.
Purification Method Antigen Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101082606, 114523, 18208, 522767, 9423
Target Species Human, Mouse, Rat, Monkey, Bovine, Feline
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.