Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547105
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547105 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547105 Supplier Invitrogen™ Supplier No. PA547105
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse Netrin-4 is observed, 15% cross-reactivity with recombinant chicken (rch) Netrin-1 is observed and less than 5% cross-reactivity with recombinant mouse (rm) Netrin-G1a, Netrin-G2a, and rchNetrin-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

NTN4 contains 3 laminin EGF-like domains, 1 laminin N-terminal domain and 1 NTR domain. NTN4 may play an important role in neural, kidney and vascular development. It promotes neurite elongation from olfactory bulb explants. NTN4 belongs to a family of proteins related to laminins (see LAMA1, MIM 150320) Koch et al. (2000) [PubMed 11038171].[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTN4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene NTN4
Gene Accession No. Q9HB63
Gene Alias Beta-netrin; CDY; CDY1A; FLJ23180; FLJ77185; hepar-derived netrin-like protein; netrin 4; netrin 4; netrin-4; netrin-4; Ntn4; PRO3091; RGD1565947
Gene Symbols NTN4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Netrin-4 Val26-Lys628.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59277
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.