Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589071 Supplier Invitrogen™ Supplier No. PA589071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSDPQCQNYC NMSDTRVHMS LRRYCQQDHV LRAQVLASEA AGPAWQRLAV RVLAVYKQRA QPVRRGDQDA WVPRADLTCG CLRLQPGTDY LLLGSAVGDP DPTRLILDRH GLALPWRPRW ARPLKRLQQE ERAGGCRGVR APTPSPRPEH; Positive Samples: Mouse kidney, Rat kidney.

NTN5 gene ontology annotations related to this gene include neurogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTN5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ntn5
Gene Accession No. Q3UQ22, Q8WTR8
Gene Alias Gm484; netrin 5; netrin-1-like; netrin-1-like protein; Netrin-5; NTN5; RGD1565197; zmp:0000000590
Gene Symbols Ntn5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 340-489 of human NTN5 (NP_6658061).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126147, 243967, 308587
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.