Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NTN5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571162
Description
This target displays homology in the following species: Cow: 100%; Dog: 92%; Guinea Pig: 92%; Human: 100%; Mouse: 77%; Pig: 100%; Rat: 77%.
NTN5 gene ontology annotations related to this gene include neurogenesis.
Specifications
NTN5 | |
Polyclonal | |
Unconjugated | |
Ntn5 | |
Gm484; netrin 5; netrin-1-like; netrin-1-like protein; Netrin-5; NTN5; RGD1565197; zmp:0000000590 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
126147 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WTR8 | |
Ntn5 | |
synthetic peptide directed towards the C-terminal region of human NTN5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction