Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTNG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566811 Supplier Invitrogen™ Supplier No. PA566811
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PYPLVWGHYDL CKTQIYTEEG KVWDYMACQP ESTDMTKYLK VKLDPPDITC GD Highest antigen sequence identity to the following orthologs - mouse 89%, rat 49%.

Netrin G1 (NTNG1) belongs to a conserved family of proteins that act as axon guidance cues during vertebrate nervous system development (Nakashiba et al., 2000).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTNG1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NTNG1
Gene Accession No. Q9Y2I2
Gene Alias A930010C08Rik; AI853992; axon guidance molecule; Kiaa0976; laminet 1; laminet-1; LMNT1; netrin G1; netrin G1f; netrin-G1; Ntng1; RGD1563465; UNQ571/PRO1133; YLSR571
Gene Symbols NTNG1
Host Species Rabbit
Immunogen Recombinant Human NTNG1. Recombinant protein control fragment (Product #RP-107469).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.