Learn More
Invitrogen™ NTNG2 Monoclonal Antibody (349724)

Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Netrin-G1a, rmNetrin-1, or rmNetrin-4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | NTNG2 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 349724 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Ntng2 |
| Gene Accession No. | Q8R4F1 |
| Gene Alias | 2610016D08Rik; bA479K20.1; bA479K20.1 (novel protein); KIAA1857; laminet 2; laminet-2; LHLL9381; Lmnt2; LOW QUALITY PROTEIN: netrin-G2; netrin G1; netrin G2; netrin-G2; NTNG1; NTNG2; UNQ9381/PRO34206 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.