Learn More
NTPCR Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305696AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
Has nucleotide phosphatase activity towards ATP, GTP, CTP, TTP and UTP. Hydrolyzes nucleoside diphosphates with lower efficiency.Specifications
NTPCR | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9BSD7 | |
NTPCR | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NTPCR | |
RP4-659I19.2, C1orf57, HCR-NTPase, RP4-678E16.2, NTPase, cancer-related nucleoside-triphosphatase, human cancer-related NTPase, nucleoside triphosphate phosphohydrolase, nucleoside-triphosphatase C1orf57 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84284 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.