Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTRK2 Mouse anti-Human, Clone:8D2E8, Abnova™

Catalog No. 89333524 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-524 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-524 Supplier Abnova Corporation Supplier No. MAB17288
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human NTRK2.

This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation. Mutations in this gene have been associated with obesity and mood disorders. Alternate transcriptional splice variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen NTRK2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8D2E8
Conjugate Unconjugated
Description neurotrophic tyrosine kinase, receptor, type 2
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene NTRK2
Gene Alias GP145-TrkB, TRKB
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 207-339 of of human NTRK2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 4915
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.