Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTRK3 Mouse anti-Human, Clone: OTI24B3, liquid, TrueMAB™
SDP

Catalog No. 501671672
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1672 Supplier OriGene Supplier No. TA500536
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRKC, a member of the insulin receptor subfamily of tyrosine protein kinases, is a receptor for neurotrophin-3 (NT-3). Known substrates for the TRK receptors are SHC, PI-3 kinase, and PLCG1. The different isoforms do not have identical signaling properties. The protein is widely expressed, mainly in the nervous tissue. The isoform B is expressed in a relatively large amount in the adult brain comparatively to fetal brain. TRKC is subject to ligand-mediated auto-phosphorylation. The protein structure contains 2 immunoglobulin-like C2-type domains and 2 leucine-rich (LRR) repeats.

Specifications

Antigen NTRK3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI24B3
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NTRK3
Gene Accession No. Q16288
Gene Alias GP145-TrkC, TRKC, gp145(trkC)
Gene Symbols NTRK3
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human NTRK3 expression vector
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4916
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.