Learn More
NTRK3 Mouse anti-Human, Clone: OTI4E10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500538

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TRKC, a member of the insulin receptor subfamily of tyrosine protein kinases, is a receptor for neurotrophin-3 (NT-3). Known substrates for the TRK receptors are SHC, PI-3 kinase, and PLCG1. The different isoforms do not have identical signaling properties. The protein is widely expressed, mainly in the nervous tissue. The isoform B is expressed in a relatively large amount in the adult brain comparatively to fetal brain. TRKC is subject to ligand-mediated auto-phosphorylation. The protein structure contains 2 immunoglobulin-like C2-type domains and 2 leucine-rich (LRR) repeats.Specifications
NTRK3 | |
Monoclonal | |
Unconjugated | |
NTRK3 | |
GP145-TrkC, TRKC, gp145(trkC) | |
Mouse | |
Affinity Chromatography | |
RUO | |
4916 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4E10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q16288 | |
NTRK3 | |
Full-length protein expressed in 293T cell transfected with human NTRK3 expression vector | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.