Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555369
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555369 Supplier Invitrogen™ Supplier No. PA555369
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DFLTNMHTSK LIQEDILDTG NDKNGKEEVI KRKIPYILKR QLYENKPRRP Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 70%.

NTS is a common precursor for two peptides, neuromedin N and neurotensin. Neurotensin is a secreted tridecapeptide, which is widely distributed throughout the central nervous system, and may function as a neurotransmitter or a neuromodulator. It may be involved in dopamine-associated pathophysiological events, in the maintenance of gut structure and function, and in the regulation of fat metabolism. Neurotensin also exhibits antimicrobial activity against bacteria and fungi. Tissue-specific processing may lead to the formation in some tissues of larger forms of neuromedin N and neurotensin. The large forms may represent more stable peptides that are also biologically active.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTS
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NTS
Gene Accession No. P30990
Gene Alias 5033428E16Rik; Large neuromedin N; Neuromedin N; neurotensin; Neurotensin/neuromedin N; NmN; NMN-125; NN; NT; NT/N; NTS; NTS1; pro-neurotensin/neuromedin; Tail peptide
Gene Symbols NTS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NTS. Recombinant protein control fragment (Product #RP-88682).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4922
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.