Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NTS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588483 Supplier Invitrogen™ Supplier No. PA588483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDSEEEMKAL EADFLTNMHT SKISKAHVPS WKMTLLNVCS LVNNLNSPAE ETGEVHEEEL VARRKLPTAL DGFSLEAMLT IYQLHKICHS RAFQHWELIQ EDILDTGNDK NGKEEVIKRK IPYIL; Positive Samples: Mouse brain, Mouse kidney; Cellular Location: Cytoplasmic vesicle, secretory vesicle.

NTS is a common precursor for two peptides, neuromedin N and neurotensin. Neurotensin is a secreted tridecapeptide, which is widely distributed throughout the central nervous system, and may function as a neurotransmitter or a neuromodulator. It may be involved in dopamine-associated pathophysiological events, in the maintenance of gut structure and function, and in the regulation of fat metabolism. Neurotensin also exhibits antimicrobial activity against bacteria and fungi. Tissue-specific processing may lead to the formation in some tissues of larger forms of neuromedin N and neurotensin. The large forms may represent more stable peptides that are also biologically active.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTS
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NTS
Gene Accession No. P20068, Q9D3P9
Gene Alias 5033428E16Rik; Large neuromedin N; Neuromedin N; neurotensin; Neurotensin/neuromedin N; NmN; NMN-125; NN; NT; NT/N; NTS; NTS1; pro-neurotensin/neuromedin; Tail peptide
Gene Symbols NTS
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299757, 67405
Target Species Mouse, Rat, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.