Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTSR1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132887
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132887 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132887 Supplier Thermo Scientific Supplier No. PA132887

Rabbit Polyclonal Antibody

PA1-32887 detects NTR1 from human samples.

TFIP11 is a nuclear speckle-localized protein that may play a role in spliceosome disassembly in Cajal bodies (Stanek et al., 2008 [PubMed 18367544]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTSR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NTSR1
Gene Accession No. P30989
Gene Alias bK445C9.6; FLJ22086; High-affinity levocabastine-insensitive neurotensin receptor; hNtr1; neurotensin receptor 1; neurotensin receptor 1 (high affinity); neurotensin receptor type 1; NT receptor-1; NT1 receptor; NT-1r; NTR; NTR1; NT-R-1; NTR-1; NTRH; NTRR; NTS1; NTS-1; Ntsr; Ntsr1; RNTR1; Spp382; TIP39
Gene Symbols NTSR1
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from C-terminus of human NTR1
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4923
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.