Learn More
NTSR2 Polyclonal Antibody, Invitrogen™
Description
PA1-32696 detects NTR2 from human samples.
Specifications
Specifications
| Antigen | NTSR2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | NTSR2 |
| Gene Accession No. | O95665 |
| Gene Alias | High-affinity levocabastine-sensitive neurotensin receptor; levocabastine-sensitive neurotensin receptor; low affinity neurotensin receptor; low-affinity levocabastine-sensitive neurotensin receptor; neurotensin receptor 2; neurotensin receptor type 2; neurotensin receptor, type 2; NT2 receptor; NT2R; NTR2; NT-R-2; NTR2 receptor; NTRL; Ntsr2 |
| Gene Symbols | NTSR2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.