Learn More
Tocris Bioscience™ nTZDpa
Catalog No. 215010R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-501-0R 10 mg
21-505-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-501-0R Supplier Tocris Bioscience™ Supplier No. 2150/10
This item is not returnable. View return policy
Potent, selective PPARγ partial agonist
nTZDpa is a potent, selective non-thiazolidinedione PPARγ partial agonist (EC50 = 57 nM); produces ∽25% maximum efficacy. Antagonizes full agonist activity by ∽60% (IC50 ∽ 285 nM). Displays no activity at PPARα or PPARδ receptors. Produces altered receptor conformation, and regulates adipocyte development and gene expression, in a differential manner to full PPARγ agonists. Modulates metabolism and insulin sensitivity without causing cardiac hypertrophy in mice in vivo.
Specifications
Quantity 10 mg
Product Type nTZDpa
Molecular Weight (g/mol) 428.33
Inhibitors nTZDpa