Learn More
Promega NUAK2 Kinase Enzyme System, 1mg
Catalog No. PRV6593 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6593 1 mg
1 options
Catalog No. PRV6593 Supplier Promega Supplier No. V6593
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human NUAK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NUAK2 or SNF1/AMP kinase-related kinase (SNARK) is a member of the NUAK family of SNF1-like kinase 2.
  • Easily Screen and Profile NUAK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C