Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUAK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582371
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582371 Supplier Invitrogen™ Supplier No. PA582371
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DPKEQKPPQA SGLLLHRKGI LKLNGKFSQT ALELAAPTTF GSLDELAPPR PLARASRPSG AVSEDSILSS ESFDQLDLPE RLPEPPLRGC VSVDNLTGLE EPPSEGPGSC LRRWRQDPLG DSCFSLTDCQ EVTATYRQAL RVCSKLT Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 81%.

NUAK2 or SNF1/AMP kinase-related kinase (SNARK) is a member of the NUAK family of SNF1-like kinase 2. NUAK2 is activated by muscle contraction and is a unique mediator of contraction-stimulated glucose transport in skeletal muscle (1). NUAK2 is involved in cellular stress responses linked to obesity and type 2 diabetes. NUAK2 shows kinase activity against a synthetic test peptide and the activity in keratinocytes is increased by AMP and 5-amino-4-imidazolecarboxamide riboside, implying that AMPK kinase-dependent pathway can activate NAUK2. Glucose deprivation also increases NAUK2 activity in baby hamster kidney fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUAK2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nuak2
Gene Accession No. Q9H093
Gene Alias 1200013B22Rik; EMBL:AAH81899.1}; mKIAA0537; NUAK family kinase 2; NUAK family SNF1-like kinase 2; NUAK family, SNF1-like kinase, 2; NUAK2; nuak2 {ECO:0000312; Omphalocele kinase 2; Omphk2; SNARK; SNF1/AMP activated protein kinase; SNF1/AMP kinase-related kinase; SNF1/AMP-activated protein kinase; UV126
Gene Symbols Nuak2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUAK2. Recombinant protein control fragment (Product #RP-89800).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81788
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.